Manganese(II) nitrate hexahydrate 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Manganous nitrate hexahydrate

L14040.14 L14040.22 L14040.36
L14040.14EA 27.9 EUR
L14040.14 L14040.22 L14040.36
Manganese(II) nitrate hexahydrate 98
Manganese (II) nitrate hexahydrate
Formula: Mn(NO₃)₂·6H₂O
MW: 287.04 g/mol
Melting Pt: 24.9
Density: 1.72 g/cm³ (30 °C)
Storage Temperature: Ambient
CAS Number: 17141-63-8
EINECS: 233-828-8
UN: 2724
ADR: 5.1 III
Merck Index: 13,05756

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR