Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, Wasser_10 99%

Supplier: Thermo Fisher Scientific

Synonyms: ATP disodium salt hydrate, 5′-ATP-Na2 ATP disodium salt hydrate, Adenosine 5'-Triphosphate Disodium Salt Hydrate, Adenosine-5'-triphosphate disodium salt hydrate, 5'-ATP 2Na Hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate disodium salt hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt xhydrate, Adenosine 5'-triphosphoric acid disodium hydrate

L14522.03 L14522.06
L14522.03EA 47.6 EUR
L14522.03 L14522.06
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, Wasser_10 99%
Adenosine 5'-triphosphate disodium salt (ATP disodium salt) hydrate

Purity: 99% CAS No.: 34369-07-8 Molecular Formula: C10H14N5Na2O13P3.xH2O Formula Weight: 551.15(anhy) Storage Temperature: -30°C to -10°C Merck Reference: 14,155 Optical Rotation: -19° (c=3 in water) MDL No.: MFCD00150755

Formula: C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
MW: 551.14 g/mol (anhy)
Storage Temperature: Freezer
MDL Number: MFCD00150755
CAS Number: 34369-07-8
EINECS: 213-579-1
Merck Index: 14,155

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR