4-Chloro-2-fluoro-6-iodoaniline 96%

Supplier: Thermo Fisher Scientific
Danger

L17658.03 L17658.06
L17658.03EA 35.3 EUR
L17658.03 L17658.06
4-Chloro-2-fluoro-6-iodoaniline 96%
4-Chloro-2-fluoro-6-iodoaniline

Purity: 96% CAS No.: 216393-67-8 Molecular Formula: C6H4ClFIN Formula Weight: 271.46 Melting Point: 39-41° UN No.: 2811 MDL No.: MFCD01318122

Formula: C₆H₄ClFIN
MW: 271.46 g/mol
Melting Pt: 39…41 °C
Storage Temperature: Ambient
MDL Number: MFCD01318122
CAS Number: 216393-67-8
UN: 2811
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR