1H-Pyrazole-4-boronic acid pinacol ester ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

L19654.03 L19654.06
L19654.03EA 83.8 EUR
L19654.03 L19654.06
1H-Pyrazole-4-boronic acid pinacol ester ≥98%
1H-Pyrazole-4-boronic acid pinacol ester

Purity: 98% CAS No.: 269410-08-4 Molecular Formula: C9H15BN2O2 Formula Weight: 194.04 Melting Point: 145-149° MDL No.: MFCD03453063

Formula: C₉H₁₅BN₂O₂
MW: 194.039 g/mol
Melting Pt: 145…149 °C
Storage Temperature: Ambient
MDL Number: MFCD03453063
CAS Number: 269410-08-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR