1,4-Benzodioxane-6-boronic acid 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2,3-Dihydro-1,4-benzodioxine-6-boronic acid

L20296.03 L20296.06
L20296.03EA 63.3 EUR
L20296.03 L20296.06
1,4-Benzodioxane-6-boronic acid 97%
1,4-Benzodioxane-6-boronic acid

Purity: 97% CAS No.: 164014-95-3 Molecular Formula: C8H9BO4 Formula Weight: 179.97 Beilstein Reference: 7478648 MDL No.: MFCD01009696

Formula: C₈H₉BO₄
MW: 179.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01009696
CAS Number: 164014-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR