Freeze dryer, console, FreeZone®
Supplier: LABCONCO
700612030 700612130 700612230 700612330 700621030 700611030 700611130 700611230 700611330 700621130 700622030 700622130 701211030 701211130 701211230 701212030 701212130 701212230 701221030 701221130 701222030 701222130 701811030 701811130 701811230 701812030 701812130 701812230 701821030 701821130 701822030 701822130 710611030 710611130 710611230 710612030 710612130 710612230 710621030 710621130 710622030 710622130 711211030 711211130 711211230 711212030 711212130 711212230 711221030 711222130 711221130 711222030 700611050 700611150 700611250 700611350 700612050 700612150 700612250 700612350 700621050 700621150 700622050 700622150 701211050 701211150 701211250 701212050 701212150 701212250 701221050 701221150 701222050 701222150 701811050 701811150 701811250 701812050 701812150 701812250 701821050 701821150 701822050 701822150 710611050 710611150 710611250 710612050 710612150 710612250 710621050 710621150 710622050 710622150 711211050 711211150 711211250 711212050 711212150 711212250 711221050 711221150 711222050 711222150 701811010
LBCP700612030EA 21980 EUR
LBCP700612030 LBCP700612130 LBCP700612230 LBCP700612330 LBCP700621030 LBCP700611030 LBCP700611130 LBCP700611230 LBCP700611330 LBCP700621130 LBCP700622030 LBCP700622130 LBCP701211030 LBCP701211130 LBCP701211230 LBCP701212030 LBCP701212130 LBCP701212230 LBCP701221030 LBCP701221130 LBCP701222030 LBCP701222130 LBCP701811030 LBCP701811130 LBCP701811230 LBCP701812030 LBCP701812130 LBCP701812230 LBCP701821030 LBCP701821130 LBCP701822030 LBCP701822130 LBCP710611030 LBCP710611130 LBCP710611230 LBCP710612030 LBCP710612130 LBCP710612230 LBCP710621030 LBCP710621130 LBCP710622030 LBCP710622130 LBCP711211030 LBCP711211130 LBCP711211230 LBCP711212030 LBCP711212130 LBCP711212230 LBCP711221030 LBCP711222130 LBCP711221130 LBCP711222030 LBCP700611050 LBCP700611150 LBCP700611250 LBCP700611350 LBCP700612050 LBCP700612150 LBCP700612250 LBCP700612350 LBCP700621050 LBCP700621150 LBCP700622050 LBCP700622150 LBCP701211050 LBCP701211150 LBCP701211250 LBCP701212050 LBCP701212150 LBCP701212250 LBCP701221050 LBCP701221150 LBCP701222050 LBCP701222150 LBCP701811050 LBCP701811150 LBCP701811250 LBCP701812050 LBCP701812150 LBCP701812250 LBCP701821050 LBCP701821150 LBCP701822050 LBCP701822150 LBCP710611050 LBCP710611150 LBCP710611250 LBCP710612050 LBCP710612150 LBCP710612250 LBCP710621050 LBCP710621150 LBCP710622050 LBCP710622150 LBCP711211050 LBCP711211150 LBCP711211250 LBCP711212050 LBCP711212150 LBCP711212250 LBCP711221050 LBCP711221150 LBCP711222050 LBCP711222150 LBCP701811010
Freeze dryer, console, FreeZone®
Freeze Dryers
FreeZone® console freeze dryers with or without stoppering tray dryers are designed for lyophilising moderate to large loads and stoppering vials or serum bottles under vacuum. The console features a front viewing window and interior space for a vacuum and accommodation for a full size tray dryer such as the FreeZone® stoppering tray dryer pump (sold separately).
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ end