Microscope slides, Snowcoat®
Supplier: LEICA HISTOLOGY
3808100GEIP 3808100GEP
LEIH3808100GEIPEA 177 EUR
LEIH3808100GEIP LEIH3808100GEP
Microscope slides, Snowcoat®
Slides Microscope Slides
These slides are designed for optimisal use with xylene-resistant pens or pencils.
  • Ground edges
  • Six colours available
  • Easy colour-coded identification