Stage without Temperature Control
Supplier: LEICA MICROSYSTEMS
11522078
LEIM11522078EA 776 EUR
LEIM11522078
Stage without Temperature Control
Microscopes
The fixed stage plate guarantee long-term stability regardless of environmental conditions.
  • High-quality aluminum
  • Ceramic-coated
  • Extremely scratchproof
  • Precisely plane-parallel
  • Three point mounting
Informace pro objednávání: The fixed stage plate is supplied with a round 88 mm insert with a 10 mm opening.