Microscope documentation cameras
Supplier: LEICA MICROSYSTEMS
13613735 11547106 11547114 11547118 11547113 11547110
630-3195EA 0 EUR
630-3195 630-3198 LEIM11547114 630-3437 630-3458 LEIM11547110
Microscope documentation cameras
Cameras
Leica (brightfield) colour cameras provide outstanding colour fidelity due to state-of-the-art colour interpolation algorithms performed in the camera head. Leica fluorescence cameras provide high sensor sensitivity allowing short exposure times and, therefore, preventing photo bleaching, so actively protecting cells from any photo damage.
  • Brightfield documentation cameras
  • Even fine structural and colour details can be distinguished due to appropriate pixel sizes for every desired microscope magnification
  • High definition (HD) display directly on a monitor