Stirrers with PTFE blade
Supplier: Lenz Laborglas GmbH & CO.KG
06315001 06315003 06315005 06315007 06315008 06315011 06315015 06315031 06315035 06315037
LENZ06315001EA 81 EUR
LENZ06315001 LENZ06315003 LENZ06315005 LENZ06315007 LENZ06315008 LENZ06315011 LENZ06315015 LENZ06315031 LENZ06315035 LENZ06315037
Stirrers with PTFE blade
Stirrers Stirring Tools
Stirrer made of precision calibrated glass tube and shaft made of DURAN® tubing with PTFE blade.
  • With ground and polished bearing surface
  • With snap-fastener for replacement of PTFE blades