Performance Professional Freezers with Static Cooling and Plastic Inner Liner, FFFsg
Supplier: Liebherr
FFFSG4001 FFFSG5501 FFFSG6501
LIEBFFFSG4001EA 2039 EUR
LIEBFFFSG4001 LIEBFFFSG5501 LIEBFFFSG6501
Performance Professional Freezers with Static Cooling and Plastic Inner Liner, FFFsg
Freezers Laboratory / General Purpose Freezers
Freezers designed for professional use. Reliably store non temperature-sensitive items even at ambient temperatures up to +40 °C with a constant cooling power. They are enormously robust, long-lasting, easy to clean and particularly sustainable, not only thanks to natural refrigerant. At the same time, they fulfil all the valid directives, standards and norms, including in terms of hygiene and HACCP concept. The efficient cooling system not only ensures low energy and overall operating cost, also safe storage of items not requiring validation of temperature stability and homogeneity.