Cyanuric acid

Supplier: Lovibond

Synonyms: 1,3,5-Triazinetrion, 1,3,5-Triazine-2,4,6-triol, 1,3,5-Triazinane-2,4,6-trione, Isocyanuric acid

511370BT 511371BT
LOVI511370BTEA 25.8 EUR
LOVI511370BT LOVI511371BT
Cyanuric acid
Cyanuric acid
Formula: C₃H₃N₃O₃
MW: 129.08 g/mol
Density: 2.5 g/cm³ (20 °C)
MDL Number: MFCD00082990
CAS Number: 108-80-5
EINECS: 203-618-0
Merck Index: 13,02728

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR