Drop test titration kits and reagent packs, MiniKit
Supplier: Lovibond
56K005501
LOVI56K005501EA 67.4 EUR
LOVI56K005501
Drop test titration kits and reagent packs, MiniKit
Environmental Test Kits
Derived from laboratory burette methods, the drop test range is an established methodology used across industry and which forms an integral part of the analytical program of many a speciality chemical supplier. The reagents are quick to use and give results fast, making the drop test the method of choice for many customers.
Delivery information: Kit is supplied with all reagents and equipment to perform the test, housed in a plastic case. Reagent pack is supplied in a carton containing test reagents and instruction sheet only.