THPS Test Kit
Supplier: Lovibond
56K701210
LOVI56K701210EA 322.5 EUR
LOVI56K701210
THPS Test Kit
Environmental Test Kits
THPS stands for Tetra Hydroxymethyl Phosphonium Sulphate. It is commonly regarded as an environmentally friendly biocide that is readily degradable and has low volatility. As a broad-spectrum biocide, it is effective against the most common types of bacteria that cause problems in cooling water, especially Sulphate Reducing Bacteria (SRB). THPS is also used for the control of Legionella. It inhibits the growth of algae, bacteria, yeasts and fungi in a wide range of industrial applications.
  • Specially for cooling water and other industrial applications
  • Effective against Sulphate Reducing Bacteria (SRB)
  • Drop tests for up to 100 tests
  • Easy to handle
With this test kit you can control the THPS concentration anytime and anywhere. Our kits are designed for mobile use directly on site. This way, you quickly have certainty about the correct dosage, because misdosing can also cause damage to the water system and the material. This will save both time and money at the same time. Our test kits also provide compliance with the applicable regulations.
Microorganisms and bacteria are among the main causes of problems for cooling water and other industrial water. In the worst case, legionella can form. THPS is one of the most commonly used nonoxidising biocides to combat these problems. Its effectiveness also depends on the correct dosage. With this test kit, you can quickly and easily monitor the use of THPS anywhere at any time.
Application: Cooling water, disinfection control.
Delivery information: Includes: Plastic case, THPS reagent 1, THPS reagent 2, THPS reagent 3, syringe plastic 20 ml, two titration jars with cap plastic 60 ml, instruction manual.