Multi-parameter meter (pH/conductivity/oxygen), handheld, SensoDirect 150
Supplier: Lovibond
724210 724200 724220
LOVI724210EA 703.6 EUR
LOVI724210 LOVI724200 LOVI724220
Multi-parameter meter (pH/conductivity/oxygen), handheld, SensoDirect 150
Electrochemistry Meters pH and Multi-Parameter Meters
The SensoDirect 150 is designed for multi purpose operation and measures pH/Redox, dissolved oxygen and conductivity/TDS.
  • Data logger function, results can be downloaded to a PC for further analysis
  • Automatic calibration + buffer recognition
  • Large LCD display, intuitive user interface, HOLD function
  • Flip out stand for bench top use
  • Rugged protective casing for field use and built-in electrode holder for safe storage
Typical applications include: Drinking water, cooling/boiler water, waste water, pool water, surface water, water treatment companies,More Product Information