Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

Synonyms: Sodium 1-((4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexane-1-carbonyl)oxy)-2,5-dioxopyrrolidine-3-sulphonate, Sulphosuccinimidyl-trans-4-(N-maleimidomethyl)cyclohexane-1-carboxylate, Sulpho-SMCC Sodium Salt, SMCC, Water-Soluble, 4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulpho-N-hydroxysuccinimide ester sodium salt, sulpho-SMCC, Sulfo-SMCC

M-Clarity™ quality level = MQ300

M6035-10MG M6035-50MG
M6035-10MGEA 135 EUR
M6035-10MG M6035-50MG
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), Sigma-Aldrich®
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)

4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt,powder (Sulfo-SMCC)

Formula: C₁₆H₁₈N₂O₉S
MW: 436.37 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00054978
CAS Number: 92921-24-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c01fe9b1cbbf7eca06a0cfa53835dd32254d01c4&vid=a69d252d6eee2a3265d683431bf6722c479af17e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) White to Light Brown
Appearance (Form) Powder
Solubility (Colour) Colorless to Light Yellow
Solubility (Turbidity)
2.5 mg/mL, H2O
Clear
Proton NMR spectra Conforms to Structure
Carbon 39.0 - 49.0 %
Nitrogen 5.5 - 7.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR