1-Methoxy-5-methylphenazinium methyl sulphate 95, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Warning

Synonyms: Methoxy-PMS, 1-Methoxy-5-methylphenazinium Methyl sulphate, 1-Methoxyphenazine methosulphate, 1-methoxy PMS

MQ200

M8640-100MG M8640-500MG
M8640-100MGEA 444 EUR
M8640-100MG M8640-500MG
1-Methoxy-5-methylphenazinium methyl sulphate 95, Sigma-Aldrich®
1-Methoxy-5-methylphenazinium methyl sulphate

1-Methoxy-5-methylphenazinium methyl sulfate (MPMS) is a photochemically stable electron-transport mediator between NAD(P)H and tetrazolium dyes. The reduced form of MPMS converts tetrazolium salt to purple precipitate formazan. It is a very stable and non-enzymic electron carrier that mediates electron transfer by dehydrogenases in vitro to various electron acceptors such as tetrazolium dyes or the electrode of an enzymic electric cell.


  • 1-Methoxy-5-methylphenazinium methyl sulfate has been used as a component of buffers or solutions for the assay of lactate dehydrogenase and muscle succinate dehydrogenase.

Formula: C₁₅H₁₆N₂O₅S
MW: 336.37 g/mol
Storage Temperature: Room Temperature
MDL Number: MFCD00040648
CAS Number: 65162-13-2
EINECS: 265-579-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55a93a4acca2726ff61a4fba2055c744fe362923&vid=603515461c72f091004889d4ca8908e9e3b94b8a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) Red to Brown
Appearance (Form) Powder
Solubility (Colour) Red
Solubility (Turbidity)
1 mg/ml, H₂O
Clear
Proton NMR spectrum Conforms to Structure
Nitrogen ≥7.9%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR