Malachite green oxalate, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Danger

Synonyms: Malachite green hemioxalate, N,N,N',N'-Tetramethyl-4,4'-diaminotriphenylcarbenium oxalate, Bis (N-{4-[[4-(dimethylamino)phenyl](phenyl)methylene]cyclohexa-2,5-dien-1-ylidene}-N-methylmethanaminium) trioxalate, Basic Green 4, N-[4-[[4-(Dimethylamino)phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-methanaminium, ethanedioate, ethanedioate(2:2:1)

MQ200

M9015-100G M9015-25G
M9015-100GEA 278 EUR
M9015-100G M9015-25G
Malachite green oxalate, Sigma-Aldrich®
Malachite green oxalate
Formula: C₅₂H₅₄N₄O₁₂
MW: 927.02 g/mol
Melting Pt: ∼159 °C
Storage Temperature: Ambient
MDL Number: MFCD00011766
CAS Number: 2437-29-8
EINECS: 219-441-7
UN: 2811
ADR: 6.1 III
Merck Index: 13,05722
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57a681278cee6e800833eed5b1933536362627b7&vid=701185d0bb9cdb0161fd8fef0638f51c2979498a&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7723ac34e7273345d2b069c0836679df388eaaf7&vid=ec3b640e950ddc753e47d460407fdd3277490d06&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR