HPLC columns, NUCLEOSHELL® HILIC
Supplier: MACHEREY-NAGEL
763332.20 763338.20 763334.20 763336.40 763334.30 763332.30 763332.40 763332.46 763334.40 763334.46 763336.20 763336.30 763336.46 763338.30
MANA763332.20EA 700 EUR
MANA763332.20 MANA763338.20 MANA763334.20 MANA763336.40 MANA763334.30 MANA763332.30 MANA763332.40 MANA763332.46 MANA763334.40 MANA763334.46 MANA763336.20 MANA763336.30 MANA763336.46 MANA763338.30
HPLC columns, NUCLEOSHELL® HILIC
Chromatography Columns
NUCLEOSHELL® HILIC is a core-shell technology based stationary phase with a covalently bonded 3-N,N-dimethylaminopropane sulphonic acid ligand.
  • Efficient core-shell silica gel for fast HPLC
  • Ammonium - sulphonic acid modification
  • Hydrophilic phase for hydrophilic interaction chromatography (HILIC)
  • Ideal for reproducible and stable chromatography of highly polar analytes
  • Very short column equilibration times
HILIC columns are especially suited for the separation of hydrophilic, polar and ionic analytes under reversed phase conditions. These analytes areMore Product Information