2,6-Dimethoxyphenylboronic acid

Supplier: Thermo Scientific

AC35922.1 AC35922.50
MAYBAC35922.1EA 0 EUR
MAYBAC35922.1 MAYBAC35922.50
2,6-Dimethoxyphenylboronic acid
2,6-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
MDL Number: MFCD01318987
CAS Number: 23112-96-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR