2-Bromo-4,6-difluoroaniline

Supplier: Thermo Scientific
Warning

Synonyms: BDFA

BTB03454.250 BTB03454.1
CHMPBTB03454.250EA 0 EUR
CHMPBTB03454.250 MAYBBTB03454.1
2-Bromo-4,6-difluoroaniline
2-Bromo-4,6-difluoroaniline
Formula: C₆H₄BrF₂N
MW: 208.01 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 42…42.5 °C
MDL Number: MFCD00009639
CAS Number: 444-14-4
EINECS: 429-430-0
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR