3-Bromo-p-anisaldehyde

Supplier: Thermo Scientific

Synonyms: 3-Bromo-4-methoxybenzaldehyde

BTB06370.1 BTB06370.10 BTB06370.50
MAYBBTB06370.1EA 0 EUR
MAYBBTB06370.1 MAYBBTB06370.10 MAYBBTB06370.50
3-Bromo-p-anisaldehyde
3-Bromo-p-anisaldehyde
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 108…110 °C (1 mmHg)
Melting Pt: 54
MDL Number: MFCD00016599
CAS Number: 34841-06-0
EINECS: 252-241-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR