4-[(3,4-Dichlorobenzyl)oxy]benzaldehyde

Supplier: Thermo Scientific

Synonyms: 4-((3,4-Dichlorobenzyl)oxy)benzaldehyde

BTB09518.250 BTB09518DA
CHMPBTB09518.250EA 0 EUR
CHMPBTB09518.250 MAYBBTB09518DA
4-[(3,4-Dichlorobenzyl)oxy]benzaldehyde
4-[(3,4-Dichlorobenzyl)oxy]benzaldehyde
Formula: C₁₄H₁₀Cl₂O₂
MW: 281.1340 g/mol
MDL Number: MFCD00173652
CAS Number: 66742-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR