2,4-Dibromothiophene

Supplier: Thermo Scientific

CC28510DA CC28510.10
MAYBCC28510DAEA 0 EUR
MAYBCC28510DA MAYBCC28510.10
2,4-Dibromothiophene
2,4-Dibromothiophene
Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: –30 °C
Density: 2.14 g/cm³ (20 °C)
MDL Number: MFCD00043889
CAS Number: 3140-92-9
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR