2-Fluoro-3-methylbenzyl bromide

Supplier: Thermo Scientific

Synonyms: 1-(Bromomethyl)-2-fluoro-3-methylbenzene

CD05588DA CD05588EA
MAYBCD05588DAEA 0 EUR
MAYBCD05588DA MAYBCD05588EA
2-Fluoro-3-methylbenzyl bromide
2-Fluoro-3-methylbenzyl bromide
Formula: C₈H₈BrF
MW: 203.05 g/mol
Boiling Pt: 105…106 °C (10 mmHg)
MDL Number: MFCD00042470
CAS Number: 151412-12-3
UN: 3265
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR