3-Chloro-4-hydroxybenzaldehyde

Supplier: Thermo Scientific

RH00193.1 RH00193.10 RH00193.50
MAYBRH00193.1EA 0 EUR
MAYBRH00193.1 MAYBRH00193.10 MAYBRH00193.50
3-Chloro-4-hydroxybenzaldehyde
3-Chloro-4-hydroxybenzaldehyde
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 251
Melting Pt: 133…134 °C
Density: 1.0404 g/cm³ (20 °C)
MDL Number: MFCD00016981
CAS Number: 2420-16-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR