3-Chloro-4-fluoroaniline

Supplier: Thermo Scientific
Warning

SB00591.1 SB00591.10 SB00591.50
MAYBSB00591.1EA 0 EUR
MAYBSB00591.1 MAYBSB00591.10 MAYBSB00591.50
3-Chloro-4-fluoroaniline
3-Chloro-4-fluoroaniline
Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 230
Melting Pt: 41…44 °C
Density: 1.226 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007767
CAS Number: 367-21-5
EINECS: 206-682-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR