4'-Fluoroacetophenone

Supplier: Thermo Scientific
Warning

Synonyms: 1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone

SB00895.1 SB00895.10 SB00895.50
MAYBSB00895.1EA 0 EUR
MAYBSB00895.1 MAYBSB00895.10 MAYBSB00895.50
4'-Fluoroacetophenone
4'-Fluoroacetophenone
Formula: C₈H₇FO
MW: 138.14 g/mol
Boiling Pt: 196
Density: 1.138 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000354
CAS Number: 403-42-9
EINECS: 206-960-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR