Cyclooctanone

Supplier: Thermo Scientific
Danger

SB01231.1 SB01231.10 SB01231.50
MAYBSB01231.1EA 0 EUR
MAYBSB01231.1 MAYBSB01231.10 MAYBSB01231.50
Cyclooctanone
Cyclooctanone
Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 196
Melting Pt: 39…41 °C
Density: 0.9581 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001754
CAS Number: 502-49-8
EINECS: 207-940-2
UN: 1759
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR