2-Fluoro-5-nitroaniline

Supplier: Thermo Scientific

SB01381.1 SB01381.10 SB01381.50
MAYBSB01381.1EA 0 EUR
MAYBSB01381.1 MAYBSB01381.10 MAYBSB01381.50
2-Fluoro-5-nitroaniline
2-Fluoro-5-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00007652
CAS Number: 369-36-8
EINECS: 206-720-3
UN: 1325
ADR: 4.1 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR