3-Chloro-4-fluorophenyl isocyanate

Supplier: Thermo Scientific

Synonyms: 2-Chloro-1-fluoro-4-isocyanatobenzene

SB01907.250 SB01907.1
CHMPSB01907.250EA 0 EUR
CHMPSB01907.250 MAYBSB01907.1
3-Chloro-4-fluorophenyl isocyanate
3-Chloro-4-fluorophenyl isocyanate
Formula: C₇H₃ClFNO
MW: 171.56 g/mol
Boiling Pt: 108…112 °C (1013 hPa)
Density: 1,377 g/cm³ (20 °C)
MDL Number: MFCD00037037
CAS Number: 50529-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR