3-Chlorothiophene

Supplier: Thermo Scientific

SB02032.10 SB02032.50
MAYBSB02032.10EA 0 EUR
MAYBSB02032.10 MAYBSB02032.50
3-Chlorothiophene
3-Chlorothiophene
Formula: C₄H₃ClS
MW: 118.59 g/mol
Boiling Pt: 133…135 °C (1013 hPa)
Density: 1.276 g/cm³ (20 °C)
MDL Number: MFCD00043887
CAS Number: 17249-80-8
UN: 1992
ADR: 3 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR