5-Chloro-2-methoxybenzonitrile

Supplier: Thermo Scientific

TL00283.50
MAYBTL00283.50EA 0 EUR
MAYBTL00283.50
5-Chloro-2-methoxybenzonitrile
5-Chloro-2-methoxybenzonitrile
Formula: C₈H₆ClNO
MW: 167.59 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 99 °C
MDL Number: MFCD00052867
CAS Number: 55877-79-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR