2-Indolinone

Supplier: Thermo Scientific

Synonyms: 2-Oxindole, Oxindole, Indolin-2-one, 2,3-Dihydro-2-indolone

TL00805DA TL00805.50
MAYBTL00805DAEA 0 EUR
MAYBTL00805DA MAYBTL00805.50
2-Indolinone
2-Indolinone
Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 122…125 °C
Density: 1.32 g/cm³ (20 °C)
MDL Number: MFCD00005711
CAS Number: 59-48-3
EINECS: 200-429-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR