Super Adjustable™ Super Erecta® shelving
Supplier: Metro International
A2448NK3 A2460NK3 A1848NK3 A2436NS A2454NS A2436NC A2442NS A1842NS A1842NC A1830NS A2460NS A1824NC A1872NS A2448-NS A1860NS A2448NC A1836NC A2136NC A2430NC A2172NC A2148NC A1848NC A1442NC A1424NC A3036NC A2442NC A2460NC A3672NC A3636NC A3060NC A2472NC A3636NS A2160NC A1430NC A1860NC A2124NC A1436NC A1448NC A2142NS A1854NC A1442NS A1460NS A3660NS A1836NS A2148NS A3048NC A1854NS
METOA2448NK3EA 248 EUR
METOA2448NK3 METOA2460NK3 METOA1848NK3 METOA2436NS METOA2454NS METOA2436NC METOA2442NS METOA1842NS METOA1842NC METOA1830NS METOA2460NS METOA1824NC METOA1872NS METOA2448-NS METOA1860NS METOA2448NC METOA1836NC METOA2136NC METOA2430NC METOA2172NC METOA2148NC METOA1848NC METOA1442NC METOA1424NC METOA3036NC METOA2442NC METOA2460NC METOA3672NC METOA3636NC METOA3060NC METOA2472NC METOA3636NS METOA2160NC METOA1430NC METOA1860NC METOA2124NC METOA1436NC METOA1448NC METOA2142NS METOA1854NC METOA1442NS METOA1460NS METOA3660NS METOA1836NS METOA2148NS METOA3048NC METOA1854NS
Super Adjustable™ Super Erecta® shelving
Furniture Shelves
Designed to work in conjunction with the entire Super Erecta® System of shelves and accessories, these Super Adjustable wire shelves have a unique corner release system which allows them to be adjusted quickly and easily without the use of tools.
  • Heavy duty construction
  • Built to save space and to stand though against years of heavy use
  • Adjust and add shelves quickly and easily to reclaimwasted space
They shelf have a sleek chrome finish that makes it perfect for dry storage areas, while the open-wire construction minimises dust accumulation and allowsMore Product Information