Masterflex® L/S® Precision Pump Tubing, Versilon™ A-60-G
Supplier: Avantor Fluid Handling
MFLX06404-13EA 83.9 EUR
MFLX06404-13 MFLX06404-14 MFLX06404-16 MFLX06404-17 MFLX06404-18 MFLX06404-25
Masterflex® L/S® Precision Pump Tubing, Versilon™ A-60-G
Tubing Peristaltic Pump Tubing
Well-suited for vacuum/pressure applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-