Hand-operated detergent, waxes, alkalis and acid transfer pump
Supplier: Avantor Fluid Handling
06433-31
MFLX06433-31EA 184 EUR
MFLX06433-31
Hand-operated detergent, waxes, alkalis and acid transfer pump
Pumps Drum/Transfer Pumps
Ideal for the janitorial market for transfering detergents.
  • Tested for transferring HCl in concentrations up to 30% at 100 °F (38 °C)
  • Self-priming, will not siphon
This hand-operated drum pump is ideal for pumping detergents, waxes, alkalis and certain acids. Constructed of solvent-welded PVC body and shaft, polyethylene (PE) hose, Viton® valves and seals, this pump offers 5 strokes/gallon.