Masterflex® I/P® Precision Pump Tubing, PharMed® BPT, Avantor®
Supplier: Avantor Fluid Handling
MFLX06508-26EA 333 EUR
MFLX06508-26 MFLX06508-73 MFLX06508-82
Masterflex® I/P® Precision Pump Tubing, PharMed® BPT, Avantor®
Tubing Peristaltic Pump Tubing
Protect your light-sensitive samples.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
Masterflex® PharMed® BPT Precision pump tubing is designed to withstand over 1More Product Information