Masterflex® Torque Tee Sanitary Clamp Securing Devices, Avantor®
Supplier: Avantor Fluid Handling
MFLX23508-03EA 331 EUR
MFLX23508-03 MFLX23508-04 MFLX23508-05 MFLX23508-06 MFLX23508-07 MFLX23508-08 MFLX61126-89 MFLX38450-85
Masterflex® Torque Tee Sanitary Clamp Securing Devices, Avantor®
Tool Sets
Ensure the security of sanitary connections with preset torque output.
  • Works with any sanitary clamp wing nut and Rubberfab's Torque-Rite® nut
  • Easily reach into tight or awkward spaces
  • Choose the appropriate torque output per the clamp manufacturer's specifications
  • Audible click alerts you when the torque has been reached
  • Ratcheting mechanism disengages for easy clamp removal and is sealed within handle for durability
  • 'T' shape creates effective leverage for fast, easy one-handed tightening
The Rubberfab Torque Tee and universal socket are designedMore Product Information