Masterflex® Sanitary Tri-Clamp® Screen Gaskets, Viton, Avantor®
Supplier: Avantor Fluid Handling
30536-06 30536-08 30536-24 30536-40 30536-64
MFLX30536-04EA 70.2 EUR
MFLX30536-04 MFLX30536-22 MFLX30536-26 MFLX30536-28 MFLX30536-42 MFLX30536-44 MFLX30536-46 MFLX30536-48 MFLX30536-62 MFLX30536-66 MFLX30536-68 MFLX30536-80 MFLX30536-82 MFLX30536-84 MFLX30536-86 MFLX30536-88 MFLX30536-06 MFLX30536-08 MFLX30536-24 MFLX30536-40 MFLX30536-64
Masterflex® Sanitary Tri-Clamp® Screen Gaskets, Viton, Avantor®
Seals Gaskets
Protect your pump, valves and other components in fluid line.
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, USDA, 3-A and cGMP criteria. FDA compliant. All gaskets are animal-derivedMore Product Information