CPC® Multi-Tube Quick-Disconnect Connectors
Supplier: Avantor Fluid Handling
31050-01 31050-02 31050-04 31050-05 31050-06 31050-02BLK 31050-05BLK 31050-01BLK 31050-04BLK 31050-06BLK 31307-88BLK 31307-93BLK 31308-09BLK 31307-75BLK 31307-96BLK 31308-06BLK 31308-25BLK
MFLX31050-01EA 34.7 EUR
MFLX31050-01 MFLX31050-02 MFLX31050-04 MFLX31050-05 MFLX31050-06 MFLX31050-02BLK MFLX31050-05BLK MFLX31050-01BLK MFLX31050-04BLK MFLX31050-06BLK MFLX31307-88BLK MFLX31307-93BLK MFLX31308-09BLK MFLX31307-75BLK MFLX31307-96BLK MFLX31308-06BLK MFLX31308-25BLK
CPC® Multi-Tube Quick-Disconnect Connectors
Fittings and Connectors
Save time - with one coupling you can connect two, six, or ten lines.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • One easy-to-use quick disconnect for multiple tubing lines
  • Multiline couplings offer quick transfers and minimize down time
These CPC® multi-tube connectors simplify the manual steps in your process for quick transfers and minimise your down time. These couplings make it easy to handle multiple tubing lines in your process that require connection and reconnection. They can disconnect multiple lines with the push of aMore Product Information