Masterflex® Adapter Fittings, Male Luer to Hose Barb, Straight, Polycarbonate, Avantor®
Supplier: Avantor Fluid Handling
MFLX45504-00EA 24 EUR
MFLX45504-00 MFLX45504-02 MFLX45504-04 MFLX45504-06 MFLX45504-07 MFLX45504-08 MFLX45504-19 MFLX45504-50 MFLX45504-40
Masterflex® Adapter Fittings, Male Luer to Hose Barb, Straight, Polycarbonate, Avantor®
Fittings and Connectors
Ensure your process integrity with full material traceability.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options