Micropump® Eagle Drive™ Pump Systems
Supplier: Avantor Fluid Handling
MFLX74215-10EA 2790 EUR
MFLX74215-10 MFLX74445-05 MFLX74445-15 MFLX74445-25
Micropump® Eagle Drive™ Pump Systems
Pumps Gear Pumps
Complete pumps include head, motor, and integrated drive.
  • Variable-speed electronic controller delivers smooth, pulseless flow
  • Static seals reduce maintenance
  • Small envelope-ideal for OEM use
Electromagnetically driven, precision-geared pumps provide smooth, accurate, pulseless flow. Torque improvements and compact size are achieved with NdFeB magnets. Control speed by using a 0 to 5 VDC input signal. Drives feature thermal and overload protection.
Choose a system with a suction shoe pump head to self compensate for wear, temperature, and pressure,More Product Information