Abaque™ Peristaltic Natural Rubber Replacement Hose
Supplier: Avantor Fluid Handling
75810-21
MFLX75810-32EA 300 EUR
MFLX75810-32 MFLX75810-43 MFLX75810-48 MFLX75810-83 MFLX75810-21 MFLX75810-64
Abaque™ Peristaltic Natural Rubber Replacement Hose
Tubing Peristaltic Pump Tubing
Highly resilient with excellent abrasion resistance and strength.
  • All listed hoses in this ad are made from Natural Rubber.
  • The hose IDs are: 10, 15, 20, 25, 32, 40, 50, 65, 80, 100 mm. Each liquid end is designed to accept one particular hose ID version.
The hose of a Peristaltic Hose pump is the only component that comes in contact with the pumped medium. Abaque engineers / manufactures their hoses from high quality compounded rubber, reinforced by layers of nylon. Once constructed, the hose is then machined with precision for efficiency, accuracy,More Product Information