Masterflex Ultrapharm™ L/S® Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
MFLX77312-00EA 5930 EUR
MFLX77312-00 MFLX77312-02 MFLX77312-10 MFLX77312-12
Masterflex Ultrapharm™ L/S® Pump Heads, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Every drop counts—integrate into automated filling systems for high accuracy dosing and dispensing.
  • Offset rollers and planetary gearing ensure dosing accuracy (±0.5%), low pulsation, and long tubing life
  • Masterflex double-Y tube sets ensure proper retention, repeatability, and accuracy
  • 250 rpm max speed; dosing volumes from <0.5 ml to >500 mL
  • Easy on tubing—planetary drive rollers reduce drag on tubing for longer tubing life and reduced risk of premature tubing failure
  • Whisper quiet operation reduces extra noise on the floor or in the lab