Masterflex® I/P® MasterSense™ Drives, Avantor®
Supplier: Avantor Fluid Handling
MFLX77421-00EA 10440 EUR
MFLX77421-00 MFLX77421-02 MFLX77421-00-EU MFLX77421-02-EU MFLX77421-00-UK MFLX77421-02-UK MFLX77421-00-IT MFLX77421-02-IT MFLX77421-00-IL MFLX77421-02-IL
Masterflex® I/P® MasterSense™ Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Versatile pumps for process applications – with an intuitive touch-screen interface and advanced connectivity.
  • Powerful, space-efficient drives will accept all I/P pump head and tubing options
  • 17,8 cm (7") capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Over-the-air firmware updates let you take full advantage of improvements in drive features and functionality
  • Three-level user access – gives you the option of controlling who can operate the pump