Screw top vials and caps, 9 mm
Supplier: MICROSOLV
9502S-3WCP
MICS9502S-3WCPEA 81.7 EUR
MICS9502S-3WCP
Screw top vials and caps, 9 mm
Vials
These autosampler vials are made of borosilicate glass.
  • With write on patch
  • Screw cap with silicone/PTFE pre-slit bonded septa
Suitable for HPLC and GC. Compatible with most Agilent™, Beckman™, CTC™, Gilson™, Shimadzu™, Thermo™, Variant™ and Waters™ autosamplers including Alliance™ and Acquity™.
Delivery information: Box includes 100 vials and caps.