Water purification system, AFS® 10E/15E
Supplier: Merck
ZAFS10E0DK ZAFS10E0EU ZAFS10E0BE ZAFS15E0EU ZAFS10E0FR ZAFS10E0GE ZAFS10E0IT ZAFS10E0SP ZAFS10E0SZ ZAFS10E0UK ZAFS15E0DK ZAFS15E0FR ZAFS15E0GE ZAFS15E0IT ZAFS15E0SP ZAFS15E0SZ ZAFS15E0UK ZAFS15E0BE
MILLZAFS10E0DKEA 10570 EUR
MILLZAFS10E0DK MILLZAFS10E0EU MILLZAFS10E0BE MILLZAFS15E0EU MILLZAFS10E0FR MILLZAFS10E0GE MILLZAFS10E0IT MILLZAFS10E0SP MILLZAFS10E0SZ MILLZAFS10E0UK MILLZAFS15E0DK MILLZAFS15E0FR MILLZAFS15E0GE MILLZAFS15E0IT MILLZAFS15E0SP MILLZAFS15E0SZ MILLZAFS15E0UK MILLZAFS15E0BE
Water purification system, AFS® 10E/15E
Water Purification Systems Pure and Ultra Pure Water Systems
Economical water system with Elix® technology, producing CLRW (Clinical Laboratory Reagent Water) quality with a dispensing flow rate to analyser up to 2 L/min. For clinical analysers with pure water needs up to 300 L daily.
  • Elix® electrodeionization (EDI) technology is combined with other complementary water purification techniques to provide consistent water quality
  • Elix® EDI technology uses permanently self-regenerating ion exchange resins to ensure predictable running costs
  • No resin cylinders to replace: Lower running costs
  • Full monitoring and