ImageXpress® HCS.ai High-Content Screening System
Supplier: Molecular Devices
5314901 5314902 5301118 5301119 5301121 IXHCSAIADV IXHCSAICF IXHCSAIWF 5306813 5306814 5315485 5315493 5315533 5315481 5302575 5315510 5315511 5315512 5315513 5315514 5315515 5315561 5315562 5088731 5088733 5090016
MLDV5314901EA 1460 EUR
MLDV5314901 MLDV5314902 MLDV5301118 MLDV5301119 MLDV5301121 MLDVIXHCSAIADV MLDVIXHCSAICF MLDVIXHCSAIWF MLDV5306813 MLDV5306814 MLDV5315485 MLDV5315493 MLDV5315533 MLDV5315481 MLDV5302575 MLDV5315510 MLDV5315511 MLDV5315512 MLDV5315513 MLDV5315514 MLDV5315515 MLDV5315561 MLDV5315562 MLDV5088731 MLDV5088733 MLDV5090016
ImageXpress® HCS.ai High-Content Screening System
Microscopes
The ImageXpress® HCS.ai High-Content Screening System captures your imagination with high-quality imaging and analysis, enabling you to acquire the data you need with ease.
  • Adapt to future lab needs with easy, cost-effective, on-site upgrades, protecting your initial investment
  • Capture exceptional detail in 2D and 3D biological models with swappable spinning disks, optimised pinhole deep tissue disks, and advanced water immersion objectives
  • Get clearer images faster with a highly sensitive 95% peak quantum efficiency sCMOS camera
  • Streamline complex