SpectraMax iD5 Injector System and Installation Service. Compatible with the SpectraMax iD5 MultiMode Detection Platform. Comes wit h 2 injectors for fast flash kinetics applications, for Absorbance, Fluorescence, and Luminescence. System accessorie 1 * 1 items
Supplier: Molecular Devices
SpectraMax iD5 Injector System and Installation Service. Compatible with the SpectraMax iD5 MultiMode Detection Platform. Comes wit h 2 injectors for fast flash kinetics applications, for Absorbance, Fluorescence, and Luminescence. System accessorie 1 * 1 items